Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Micro particles based on polymethacrylate, analytical standard, Supelco®

Micro particles based on polymethacrylate, analytical standard, Supelco®

1 µm, Particle suspension 10% solids, std dev <0,2 µm, Analytical standard, 5 mL
Catalog # 90875-5ML-F
Supplier:   Merck
Micro particles based on polymethacrylate, analytical standard, Supelco®
Catalog # 90875-5ML-F
Supplier:  Merck
CAS Number:  
Micro particles based on polymethacrylate, analytical standard, Supelco®

1 µm, Particle suspension 10% solids, std dev <0,2 µm, Analytical standard, 5 mL

CAS Number:  
Price
297,16 €
Log in for availability
1 * 5 mL
Product Details & Documents

The exact values for particle size and standard deviation are determined for each lot and may be found on the certificate of analysis. These values are determined with an accuracy of 0,01 μm (Coulter Multisizer). Polymethacrylate based particles have a narrow size distribution and spherical shape. They are stable at temperatures of up to 200 °C.

Documents

Specifications

Specifications

  • Particle Size:
    1 µm
  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • Description:
    Particle suspension 10% solids
  • Accuracy:
    std dev <0,2 µm
  • Cat. No.:
    90875-5ML-F
  • M-Clarity quality level:
    MQ100

Specifications