CHANNEL SEDIMENT - TRACE ELEMENTS 1 * 40 g
Catalog # PROOBCR-320R
Supplier: PROMOCHEM
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck