Accessories for multi-mode Varioskan™ reader
Luminometric emission filter, 425 nm HBW50 D25 mm
Catalog # LABLF42550
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Accessories for multi-mode Varioskan™ reader
Catalog # LABLF42550
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Jump to:
Product Details & Documents
Specifications
- Description:Luminometric emission filter, 425 nm HBW50 D25 mm
- Seq:12