Order Entry
Slovakia
Orders LinkContactUsLinkComponent
(±)-2-Bromopentane tech. 90%, Technical Grade
Glass bottle for solids, 100 g
  A17205.22
(±)-2-Bromopentane tech. 90%, Technical Grade
  A17205.22
 :  107-81-3
(±)-2-Bromopentane tech. 90%, Technical Grade
(±)-2-Bromopentane tech. 90%, Technical Grade
Glass bottle for solids, 100 g
 :  107-81-3
135,22 €
1 * 100 g
Purity: 90% CAS No.: 107-81-3 Molecular Formula: C5H11Br Formula Weight: 151.05 Flash Point: 20°(68°F) Refractive Index: 1.4403 Odour: Characteristic Melting Point: -96° Boiling Point: 117-118° Beilstein Reference: 1718829 UN No.: 2343 MDL No.: MFCD00000160
 :  (±)-2-Pentyl bromide
  •  :107-81-3
  •  :H₃CBrCH(CH₂)₂CH₃
  •  :MFCD00000160
  •  :151.05 g/mol
  •  :117…118 °C
  •  :–96 °C
  •  :1.220
  •  :20 °C (68 °F)
  •  :Ambient
  •  :203-521-3
  •  :2343
  •  :3,II
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g