Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Oleic acid

Oleic acid

Glass bottle for solids, 500 mg
Catalog # CAYM90260-500
Supplier:   CAYMAN
Oleic acid
Catalog # CAYM90260-500
Supplier:  CAYMAN
CAS Number:  112-80-1
Oleic acid
Oleic acid

Glass bottle for solids, 500 mg

CAS Number:  112-80-1
Price
35,39 €
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • MDL Number:MFCD00064242
  • Molecular Weight:282.47 g/mol
  • Merck Index:13,06898
  • Boiling Pt:286 °C (1013 hPa)
  • Melting Pt:16.7 °C
  • Density:0.8946 g/cm³ (20 °C)
  • Flash Pt:180 °C
  • Storage Temperature:Freezer
  • EINECS:204-007-1

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.