BUFFER SOLUTION PH 4.00 1 * 900 mL
Catalog # METR6.2307.200
Supplier: METROHM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffer solutions, certified (DKD)
Supplier: METTLER TOLEDO
pH buffer solutions
Supplier: METTLER TOLEDO
Potassium iodide saturated solution in water Reag. Ph. Eur. 1070504
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Lithium standard for IC, TraceCERT®, Supelco®
Supplier: Merck
PENICILLAMINE 1 * 500 mg
Supplier: Merck
Reagents for chloride determination using Lovibond® MD photometers
Supplier: TINTOMETER
BUFFER SOLUTION PH 7.00 1 * 900 mL
Supplier: METROHM