Fenazaquin certified 1 * 0,1 g
Catalog # EHERC13441000
Supplier: EHRENSTORFER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck