Acetophenone 1 * 1 mL
Catalog # EHERC10022000
Supplier: EHRENSTORFER
Frequently Bought Together
CASTICIN 1 * 10 mg
Supplier: Merck
Quizalofop (free acid) D3 (3,3,3-D3) 100 ng/µl 1 * 1,1 mL
Supplier: EHRENSTORFER
Zoxamide certified 1 * 0,1 g
Supplier: EHRENSTORFER
Testosterone, analytical standard, VETRANAL™, Supelco®
Supplier: Merck
Turbidity standards for 2100 AN and 2100 AN IS
Supplier: HACH
VWR®, Sulphur, Certified Reference Materials, in synthetic diesel
Supplier: VWR Chemicals
Accessories for ion selective meters, LAQUAtwin
Supplier: HORIBA
Buffer solutions
Supplier: REAGECON
OEKO-TEX PAH Mixture
Supplier: EHRENSTORFER
Buffer Solutions
Supplier: Thermo Fisher Scientific
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
MOGROSIDE V 1 * 200 mg
Supplier: Merck
AMMONIUM IONIC STRENGTH ADJUSTOR SOLN 1 * 500 mL
Supplier: YSI ENVIRONMENTAL