Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MCPA-butoxyethyl ester certified 1 * 0,1 g
EHERC14762000
:
EHRENSTORFER
MCPA-butoxyethyl ester certified 1 * 0,1 g
EHERC14762000
:
EHRENSTORFER
Frequently Bought Together
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Magnesium standard solution, 1000 mg/L Mg in 0,5 M nitric acid (from Mg(NO₃)₂) Certipur® standard for AAS, Supelco®
: Merck
Buffer solutions in sachets
: METTLER TOLEDO
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for acidimetry, traceable to NIST Standard Reference Material (SRM), Supelco®
: Merck
Buffer solutions, Certipur®, ready to use, 1 litre bottles, Supelco®
: Merck
pH buffers, ready to use
: WTW
VWR®, Chloride 1,000 mg/L in Water standard for ion chromatography
: VWR Chemicals
pH buffer solutions, colour coded
: HACH
Potassium Standard for IC, TraceCERT®, Supelco®
: Merck
pH Standard Solutions, IUPAC, Certified
: HACH