TLC polyester sheets
POLYGRAM® TLC polyester sheets Alox N/UV254, Aluminium oxide (Alox), 40×80 mm
Catalog # 552-0278
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Documents
Thin Layer Chromatography.pdf
Additional Product Details
Specifications
- Coating material:Aluminium oxide (Alox)
- Plate size:40×80 mm
- Description:POLYGRAM® TLC polyester sheets Alox N/UV254
- Seq:02
- Fluorescent indicator:F254