Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Accessories for chromatography systems
Accessories for chromatography systems
O-ring 11,3×2,4 mm, fluor carbon rubber (FPM), max. operating pressure 40 bar, 5
Catalog # 19-7584-01
Supplier:   Cytiva
Accessories for chromatography systems
Catalog # 19-7584-01
Supplier:  Cytiva
CAS Number:  
Accessories for chromatography systems
Accessories for chromatography systems
O-ring 11,3×2,4 mm, fluor carbon rubber (FPM), max. operating pressure 40 bar, 5
CAS Number:  
Price
95,36 €
Log in for availability
1 * 5 items
Specifications
Specifications
  • Pk:
    5
  • Description:
    O-ring 11,3×2,4 mm, fluor carbon rubber (FPM), max. operating pressure 40 bar