Methabenzthiazuron 10 ng/µl 1 * 10 mL
Catalog # EHERL14960000AL
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: METTLER TOLEDO
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: METTLER TOLEDO
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Metalaxyl 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
pH Standard Solutions, IUPAC, Certified
Supplier: HACH