Chloroform ≥98% stabilised, TECHNICAL
Plastic bottle, fluorinated, 5 L
Supplier: VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
Catalog # 22720.360
Supplier: VWR Chemicals
CAS Number: 67-66-3
Jump to:
Product Details & Documents
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
- CAS Number:67-66-3
- Formula:CHCl₃
- MDL Number:MFCD00000826
- Molecular Weight:119.38 g/mol
- Merck Index:13,02160
- Boiling Pt:61.7 °C (1013 hPa)
- Melting Pt:–63 °C
- Density:1.4832 g/cm³ (20 °C)
- EINECS:200-663-8
- ADR:6.1,III
- :Stabilised with ethanol 0.6 %
- Storage Temperature:Ambient
- UN:1888
- REACH:01-2119486657-20
Documents
Certificates
Search by Lot Number30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
P20_VWRC22720.360_SPEC_EN.pdf P20_VWRC22720.462_SPEC_EN.pdf P20_VWRC22720.550_SPEC_EN.pdf spec_VWRC22720.360 spec_VWRC22720.360
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:Min. 98 %
- Density (20/4):1.473 - 1.489
- Ethanol:0.1 - 1.0 %
Specifications
- Pack type:Plastic bottle, fluorinated
- Pk:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



