Bismuth(III) hydroxide, tech.
Glass bottle for solids, 1 kg
Supplier: Thermo Fisher Scientific
Bismuth(III) hydroxide, tech.
Catalog # 40397.A1
Supplier: Thermo Fisher Scientific
CAS Number: 10361-43-0
Jump to:
Product Details & Documents
CAS No.: 10361-43-0 Molecular Formula: Bi(OH)3 Formula Weight: 260.00 Physical Form: Amorphous Powder Odour: Odourless Solubility: Insoluble in water. Soluble in acids MDL No.: MFCD00046188
Hydrolysis of ribonucleic acid, absorbent
Synonyms: Bismuth hydroxide
- CAS Number:10361-43-0
- Formula:H₃BiO₃
- MDL Number:MFCD00046188
- Molecular Weight:260 g/mol
- Storage Temperature:Ambient
- EINECS:233-790-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



