Bismuth(III) hydroxide, tech.
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Bismuth(III) hydroxide, tech.
Catalog # 40397.30
Supplier: Thermo Fisher Scientific
CAS Number: 10361-43-0
Jump to:
Product Details & Documents
CAS No.: 10361-43-0 Molecular Formula: Bi(OH)3 Formula Weight: 260.00 Physical Form: Amorphous Powder Odour: Odourless Solubility: Insoluble in water. Soluble in acids MDL No.: MFCD00046188
Hydrolysis of ribonucleic acid, absorbent
Synonyms: Bismuth hydroxide
- CAS Number:10361-43-0
- Formula:H₃BiO₃
- MDL Number:MFCD00046188
- Molecular Weight:260 g/mol
- Storage Temperature:Ambient
- EINECS:233-790-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



