Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Tellurium dioxide, tech.

Tellurium dioxide, tech.

Plastic bottle for solids, 50 g
Catalog # 10608.18
Health
Tellurium dioxide, tech.
Catalog # 10608.18
CAS Number:  7446-07-3
Tellurium dioxide, tech.
Tellurium dioxide, tech.

Plastic bottle for solids, 50 g

CAS Number:  7446-07-3
Price
184,55 €
Log in for availability
1 * 50 g
Product Details & Documents

CAS No.: 7446-07-3 Molecular Formula: TeO2 Formula Weight: 159.60 Physical Form: Powder Density: 5.8 Odour: Odourless Merck Reference: 14,9123 MDL No.: MFCD00011263

  • CAS Number:7446-07-3
  • Formula:O₂Te
  • MDL Number:MFCD00011263
  • Molecular Weight:159.6 g/mol
  • Merck Index:14,9123
  • Boiling Pt:1245 °C (1013 hPa)
  • Melting Pt:733 °C
  • Density:5.8
  • Storage Temperature:Ambient
  • EINECS:231-193-1
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    50 g

Specifications