Inoculating needles and loops, Nunc™
Loops, 1 µl, Transparent, 50/sleeve
Catalog # 734-2138
Supplier: Thermo Fisher Scientific
Inoculating needles and loops, Nunc™
Catalog # 734-2138
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS, sterile. Disposable, problem free inoculation with smooth loop surface.
- Meet semi-quantitative standard for handling samples
- Surface treated for enhanced droplet adhesion
- Non toxic
Specifications
- Packed:50/sleeve
- Capacity:1 µl
- Colour:Transparent
- Description:Loops
- Seq:02
- Cat. No.:734-2138
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact