Order Entry
Slovakia
ContactUsLinkComponent
Magnetic valve extention set for waste line VZ 7162. For aggressive media! Attention: For each measuring position >1 one of these se ts is required. Scope of delivery: reqired valve support for 4 magnetic valves, incl. connectors and cable to ViscoPu 1 * 1 items
Catalog # SCOI285421676
Supplier:  SI ANALYTICS
Magnetic valve extention set for waste line VZ 7162. For aggressive media! Attention: For each measuring position >1 one of these se ts is required. Scope of delivery: reqired valve support for 4 magnetic valves, incl. connectors and cable to ViscoPu 1 * 1 items
Catalog # SCOI285421676
Supplier:  SI ANALYTICS
Frequently Bought Together