Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

TLC plates, polyamide-6, polyester sheets, POLYGRAM®

POLYAMIDE-6 UV254, Polyamide 6, 200×200 mm, 100 µm
Catalog # 552-0153
Supplier:   MACHEREY-NAGEL
TLC plates, polyamide-6, polyester sheets, POLYGRAM®
Catalog # 552-0153
Supplier:  MACHEREY-NAGEL
TLC plates, polyamide-6, polyester sheets, POLYGRAM®
TLC plates, polyamide-6, polyester sheets, POLYGRAM®

POLYAMIDE-6 UV254, Polyamide 6, 200×200 mm, 100 µm

Price
280,31 €
Log in for availability
1 * 25 items
Product Details & Documents

Polyamide 6 layers are available as POLYGRAM® polyester sheets with or without fluorescent indicator. The separation mechanism based on hydrogen bonds to amide groups of the polymer matrix as well as on ionic, dipol and electron donor/acceptor interactions.

  • Alternative selectivity to silica 60

Application includes natural compounds, phenols, carboxylic acids, aromatic nitro compounds and especially amino acids.

Polyamide 6: ε-polycaprolactam.

Documents

Specifications
  • Coating material:
    Polyamide 6
  • Layer thickness:
    100 µm
  • Plate size:
    200×200 mm
  • Description:
    POLYAMIDE-6 UV254
  • Seq:
    04
  • Fluorescent indicator:
    F254
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.