Naphthalene 99%
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:91-20-3
- Formula:C₁₀H₈
- MDL Number:MFCD00001742
- Molecular Weight:128.17 g/mol
- Merck Index:12,06457
- Boiling Pt:218 °C (760 mmHg)
- Melting Pt:79…82 °C
- Density:0.99 g/cm³
- Flash Pt:78 °C
- Storage Temperature:Ambient
- EINECS:202-049-5
- UN:1334
- ADR:4.1,III
Documents
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals or flakes or tablets or balls
- Infrared spectrum:Conforms
- Melting point:79 to 82 °C
- GC:≥98.5 %
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



