2-Hydroxy-1-naphthaldehyde tech.
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
2-Hydroxy-1-naphthaldehyde tech.
Catalog # ACRO121605000
Supplier: Thermo Fisher Scientific
CAS Number: 708-06-5
Jump to:
Product Details & Documents
Synonyms: 2-Hydroxy-α-naphthaldehyde
- CAS Number:708-06-5
- Formula:C₁₁H₈O₂
- MDL Number:MFCD00004005
- Molecular Weight:172.18 g/mol
- Boiling Pt:192 °C (27 mmHg)
- Melting Pt:>77 °C
- Storage Temperature:Ambient
- EINECS:211-902-0
- UN:3077
- ADR:9,III
Specifications
Specification Test Results
- Appearance:Yellow to brown Moist powder
- Infrared spectrum:Conforms
- Melting point:≥77 °C (on dry basis)
- Water:25 to 60 % (K.F.)
- HPLC:≥93 % (on dry basis)
Specifications
- Pack type:Plastic bottle
- Pk:500 g



