2-Hydroxy-1-naphthaldehyde
Bottle
Catalog # APOSOR8817-25G
New Product
Supplier: Apollo Scientific
2-Hydroxy-1-naphthaldehyde
Catalog # APOSOR8817-25G
Supplier: Apollo Scientific
New Product
CAS Number: 708-06-5
Jump to:
Product Details & Documents
Synonyms: 2-Hydroxy-α-naphthaldehyde
- CAS Number:708-06-5
- Formula:C₁₁H₈O₂
- MDL Number:MFCD00004005
- Molecular Weight:172.18 g/mol
- Boiling Pt:191…193 °C/27mmHg
- Melting Pt:76…80 °C
- Storage Temperature:Ambient
- EINECS:211-902-0
- UN:3077
- ADR:9+EHS,III
Specifications
Specifications
- Pack type:Bottle



