Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

2-Hydroxy-1-naphthaldehyde

Bottle
Catalog # APOSOR8817-5G
Irritant
Environment
New Product
Supplier:   Apollo Scientific
2-Hydroxy-1-naphthaldehyde
Catalog # APOSOR8817-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  708-06-5
2-Hydroxy-1-naphthaldehyde

Bottle

CAS Number:  708-06-5
Price
25,28 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Hydroxy-α-naphthaldehyde
  • CAS Number:708-06-5
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00004005
  • Molecular Weight:172.18 g/mol
  • Boiling Pt:191…193 °C/27mmHg
  • Melting Pt:76…80 °C
  • Storage Temperature:Ambient
  • EINECS:211-902-0
  • UN:3077
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.