Order Entry
Slovakia
Orders LinkContactUsLinkComponent
ZEN 3.12 Software for Microscopy Workstation
ZEN 3.12 Software for Microscopy Workstation
ZEN 3.12 software
Catalog # ZEIS4101350002312
New Product
Supplier:   ZEISS
ZEN 3.12 Software for Microscopy Workstation
Catalog # ZEIS4101350002312
Supplier:  ZEISS
New Product
ZEN 3.12 Software for Microscopy Workstation
ZEN 3.12 software
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
ZEN 3.12 image acquisition and processing under Windows 10/Windows 11×64 user interface configurable, control of the ZEISS microscope systems and components, extensive acquisition and analysis, CZI image format.
The following functions are included: Basic 3D viewing; Colocalisation; Connect basic; Direct processing; Manual EDF; Measurement; Panorama; Spectral unmixing; ZEN Data Storage Client; Acquisition base (time series and multichannel); ZEN API Gateway includes a docker desktop license required by selected ZEN toolkits.
Compatible with the below microscope models:
Axio Imager.Z1m for Particle analysis, Axio Examiner.A1, Axio Examiner.D1, Axio Examiner.Z1, Axio Observer.A1 Entry, Axio Observer.D1 Entry, Axio Observer.Z1 High End, Axio Observer.D1 Mid Range, Axio Observer.Z1 Mid Range, Axio Imager.Z1 + ApoTome, Axio Observer.D1 + ApoTome, Axio Observer.Z1 Cell Observer SD, Axio Observer.Z1 TIRF 3, Axio Imager.A2, Axio Imager.A2m, Axio Imager.D2, Axio Imager.D2m, Axio Imager.M2, Axio Imager.M2m, Axio Imager.Z2, Axio Imager.Z2m, Axio Observer.D1 TIRF 3, Axio Imager Vario, Axio Zoom.V16, Axio Examiner.D1 Cell Observer SD, Axio Examiner.Z1 Cell Observer SD, Axio Scan.Z1, Axio Observer 3 materials, Axio Observer 5 materials, Axio Observer 7 materials, Axio Observer 3, Axio Observer 5, Axio Observer 7, Celldiscoverer 7, Axio Observer 7 Cell Observer SD, LSM 800, LSM 880, LSM 880 NLO, ELYRA 7, Lightsheet, ZEN Connect, Axioscope 5 MAT, Axioscope 5 Pol, Axioscope vario, LSM 900, LSM 980, Axiolab 5, Axiolab 5 MAT, Axiolab 5 Pol, Axioscope 5, Cell Observer SD, Lightsheet 7, Axioscan 7, Axiovert 5, Axiovert 5 digital, Axiovert 7, Lattice SIM 3, Lattice SIM 5, Lattice Lightsheet 7, LSM 910, LSM 990, Axio Imager.A1, Axio Imager.A1m, Axio Imager.D1, Axio Imager.D1 for epi-fluorescence with ApoTome equipment, Axio Imager.D1m, Axio Imager.M1, Axio Imager.M1 for KS ELISPOT, Axio Imager.M1m, Axio Imager.Z1, Axio Imager.Z1m, Axio Imager.M1m for Particle analysis, Axio Observer.A1, Axio Observer.D1, Axio Observer.D1m, Axio Observer.Z1, Axio Observer.Z1 for epi-fluorescence with ApoTome equipment, Axio Observer.Z1 Cell Observer, Axio Observer.Z1m, Axio Observer.Z1m ACR, Axio Observer.A1m, Axiophot 2, Axioplan 2 Imaging, Axiovert 200, Axiovert 200 M, Axiovert 200 M MAT, Axiovert 200 MAT, Axiovert 200 for epi-fluorescence with ApoTome equipment, Axiovert 200 M for epi-fluorescence with ApoTome equipment
Specifications
  • Description:
    ZEN 3.12 software