Pyridine ≥99.5%, AR® analytical reagent
Amber glass bottle
Supplier: MACRON AVANTOR BRAND
Pyridine ≥99.5%, AR® analytical reagent
Catalog # 6260-25
Supplier: MACRON AVANTOR BRAND
CAS Number: 110-86-1
Jump to:
Product Details & Documents
: DO NOT handle, store or open near an open flame, sources of heat or sources of ignition. Protect material from direct sunlight. For Laboratory, Research or Manufacturing use.
Synonyms: Azabenzene, Azine
- CAS Number:110-86-1
- Formula:C₅H₅N
- Molecular Weight:79,1 g/mol
- Storage Temperature:Ambient
- EINECS:203-809-9
- UN:1282
- REACH:01-2119493105-40-XXXX
Specifications
Specification Test Results
- Assay (GC):min. 99,5%
- Water (KF):max. 0,1%
- Residue after Evaporation:max. 0,005%
Specifications
- Pack type:Amber glass bottle



