Order Entry
Slovakia
Orders LinkContactUsLinkComponent
SNAP NO donor

SNAP NO donor

Supplier:   ABCAM
SNAP NO donor
Supplier:  ABCAM
Certificates

2 Options Available Below

Product Details & Documents

NO donor. Vasodilator in vitro and in vivo, that does not induce pharmacological tolerance.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  S-Nitroso-N-acetyl-D,L-penicillamine, 2-acetamido-3-methyl-3-(nitrososulphanyl)butanoic acid
  • CAS Number:67776-06-1
  • Molecular Weight:220,25 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% S-Nitroso-N-acetyl-DL-penicillamine
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 102,75 €
    102,75 €
    1 * 10 mg
       102,75 €

  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 280,72 €
    280,72 €
    1 * 50 mg
       280,72 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.