Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Human eIF4E ELISA Kit

Human eIF4E ELISA Kit

Human eIF4E ELISA Kit, 96 tests
Catalog # ABCAAB214564-96
Supplier:   ABCAM
Human eIF4E ELISA Kit
Catalog # ABCAAB214564-96
Supplier:  ABCAM
CAS Number:  
Human eIF4E ELISA Kit
Human eIF4E ELISA Kit

Human eIF4E ELISA Kit, 96 tests

CAS Number:  
Price
836,42 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human eIF4E ELISA kit is a single-wash 90-min Simplestep used to quantify Human eIF4E with a sensitivity of 79.4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human eIF4E ELISA Kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Cell culture extracts, Tissue Extracts
  • Detection range:
    109,4 - 7000 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 hr 30 min
  • Cat. No.:
    ABCAAB214564-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    79,4 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    eIF4E
  • Regulatory status:
    RUO

Specifications