Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, Decabromodiphenyl ether (BDE No. 209) 100 µg/ml in acetone, Dr Ehrenstorfer
Organic reference standard, Decabromodiphenyl ether (BDE No. 209) 100 µg/ml in acetone, Dr Ehrenstorfer
Catalog # EHERXA15898209AC
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Decabromodiphenyl ether (BDE No. 209) 100 µg/ml in acetone, Dr Ehrenstorfer
Catalog # EHERXA15898209AC
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    1163-19-5
  • Description:
    Decabromodiphenyl ether (BDE No. 209) 100 µg/ml in acetone
  • Formula:
    C₁₂Br₁₀O
  • Description:
    Decabromodiphenyl ether (BDE No. 209)

Specifications

About this item