Order Entry
Slovakia
ContactUsLinkComponent
 
Cyanogen bromide 5 mol/l in acetonitrile, Sigma-Aldrich®
Catalog # SIAL261610-100ML
Supplier:  Merck
CAS Number:  
Cyanogen bromide 5 mol/l in acetonitrile, Sigma-Aldrich®
Catalog # SIAL261610-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :105.92 g/mol
EINECS:208-051-2
UN:3286
ADR:3,I
CAS Number:506-68-3
Formula:CBrN
MDL Number:MFCD00011597