Zadaní objednávky
Slovakia
ContactUsLinkComponent
Yeast invertase
Yeast invertase
Catalog # MEGAE-INVRT
Supplier:  MEGAZYME
CAS Number:  
Yeast invertase
Catalog # MEGAE-INVRT
Supplier:  MEGAZYME
CAS Number:  

Specifications

  • Pk:
    100 mL
  • CAS No.:
    9001-57-4
  • Species:
    Yeast
  • Enzyme name:
    Invertase
  • Enzyme activity:
    ~300 U/mg protein
  • Concentration:
    ~2,000 U/ml
  • Formulation:
    In 50% (v/v) glycerol

Specifications

About this item