

Ethyl-4,4,4-trifluorocrotonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25597-16-4
:406-10-0
Formula:C₆H₇F₃O₂
MDL Number:MFCD00009903
Boiling Pt:113…115 °C
Density:1.125
Flash Pt:25 °C (77 °F)
UN:3272
ADR:3,III
:168.12 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together