Chloroacetic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:79-11-8
:
Formula:ClCH₂COOH
MDL Number:MFCD00002683
:94.5 g/mol
Merck Index:14,02112
Boiling Pt:189°C
Melting Pt:56…57 °C
Density:1,4043 g/cm³ (20 °C)
Flash Pt:126 °C
Storage Temperature:Ambient
EINECS:201-178-4
UN:1751
ADR:6.1,II
:GHS05
:GHS06
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:62.0 to 66.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together