Order Entry
Slovakia
Orders LinkContactUsLinkComponent
(±)-7-HDoHE

(±)-7-HDoHE

Supplier:   CAYMAN
(±)-7-HDoHE
Supplier:  CAYMAN
Certificates

4 Options Available Below

Product Details & Documents
  • CAS Number:90780-55-5
  • Formula:C₂₂H₃₂O₃
  • UN:1170
  • ADR:3,II
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 25 µG
    Log in for availability
    Log in for availability
    1 * 25 µG
    1 * 25 µG - 138,10 €
    138,10 €
    1 * 25 µG
       138,10 €

  • 50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG - 259,35 €
    259,35 €
    1 * 50 µG
       259,35 €

  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 491,99 €
    491,99 €
    1 * 100 µG
       491,99 €

  • 250 µG
    Log in for availability
    Log in for availability
    1 * 250 µG
    1 * 250 µG - 1 087,14 €
    1 087,14 €
    1 * 250 µG
       1 087,14 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.