

Tetrakis(dimethylamido)titanium(IV) ≥99.999%, Sigma-Aldrich®
SIAL669008-25G
: Merck
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance (Color):Light Yellow to Dark Yellow and Light
- Appearance (Form):Liquid
- Proton NMR Spectrum:Conforms to Structure
- Chloride by Ion Chromatography:< 10 ppm
- Purity:Meets Requirements 99.999% Based On Trace Metals Analysis
- Trace Metal Analysis:< 20.0 ppm
:
:4.3,II
:M-Clarity™ quality level = MQ100
:–30 °C (closed cup)
:224.17 g/mol
:221-904-3
:3129
:GHS02
:GHS05
:3275-24-9
:C₈H₂₄N₄Ti
:MFCD00014861