[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid), Sigma-Aldrich®
Catalog # SIAL407003-250ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:178.1831 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:240-162-1
UN:3265
CAS Number:16024-58-1
Formula:C₇H₁₄O₅
MDL Number:MFCD00044098