Bromoethylene 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:593-60-2
:
Formula:C₂H₃Br
MDL Number:MFCD00000183
:106.95 g/mol
ADR:3,I
:M-Clarity™ quality level = MQ100
EINECS:209-800-6
UN:1993
Frequently Bought Together