Order Entry
Slovakia
ContactUsLinkComponent
1,4-Dichlorophthalazine ≥98%
1,4-Dichlorophthalazine ≥98%
Catalog # H33295.06
CAS Number:  
1,4-Dichlorophthalazine ≥98%
Catalog # H33295.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :199.04 g/mol
Melting Pt:164…166 °C
EINECS:225-275-6
CAS Number:4752-10-7
Formula:C₈H₄Cl₂N₂
MDL Number:MFCD00006909