Sudan black B, high purity biological stain
:4197-25-5
:
:C₂₉H₂₄N₆
:MFCD00006919
:456.55 g/mol
:120…124 °C
:Ambient
:224-087-1
- Appearance:Dark brown to black Crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
Frequently Bought Together