Supelco® Organic Reference Standard, Sodium 2-mercaptoethanesulfonate
: Merck
: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338
- CAS No.:19767-45-4
- Storage conditions:RT
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00007535
- M-Clarity quality level:MQ200
- Synonym(s):2-Mercaptoethanesulfonic acid sodium salt; Coenzyme M sodium salt; HS-CoM Na; MESNA
- Formula:HSCH₂CH₂SO₃Na
- Molecular weight:164,18
Frequently Bought Together