Order Entry
Slovakia
ContactUsLinkComponent
o-Cresolphthalein complexone, Sigma-Aldrich®
o-Cresolphthalein complexone, Sigma-Aldrich®
Catalog # SIALP5631-1G
Supplier:  Merck
CAS Number:  
o-Cresolphthalein complexone, Sigma-Aldrich®
Catalog # SIALP5631-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :636.62 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:219-318-8
CAS Number:2411-89-4
Formula:C₃₂H₃₂N₂O₁₂
MDL Number:MFCD00005911