Silicon dioxide
Glass bottle for solids, 100 g
Catalog # ABCRAB255289.0100
Supplier: abcr
Prejsť na:
Product Details & Documents
Synonymá: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- Číslo CAS:7631-86-9
- Vzorec:SiO₂
- MDL číslo:MFCD00011232
- Molecular Weight:60.08 g/mol
- Merck Index:14,08493
- Bod varu:2230 °C (1013 hPa)
- Bod tavenia:1719 °C
- Hustota:2.2 g/cm³ (20 °C)
- Teplota pre skladovanie:Ambient
- EINECS:231-545-4
Špecifikácie
Špecifikácie
- Pack type:Glass bottle for solids
- Pk:100 g
Špecifikácie
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



