Order Entry
Slovakia
ContactUsLinkComponent
Bovine alpha-Chymotrypsin (from Pancreas)
Bovine alpha-Chymotrypsin (from Pancreas)
Catalog # PROOFIP6
Supplier:  PROMOCHEM
CAS Number:  
Bovine alpha-Chymotrypsin (from Pancreas)
Catalog # PROOFIP6
Supplier:  PROMOCHEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • CAS No.:
    9004-07-3
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00130481
  • Enzyme name:
    alpha-Chymotrypsin

Specifications

About this item