Chlortetracycline hydrochloride, Supelco®
Catalog # 46133-250MG-R
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:515.34 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-591-7
:GHS06
:GHS08
Storage Temperature:Refrigerator
CAS Number:64-72-2
Formula:C₂₂H₂₃ClN₂O₈·HCl
MDL Number:MFCD26142699