Supelco® PESTANAL® Organic Reference Standard, Bifenthrin
34314-100MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard, mixture of isomers
- CAS No.:99267-18-2
- Description:Bifenthrin
- Storage conditions:2...30 °C
- Hazard class description:H300 - H317 - H331 - H351 - H372 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic acid (2-methylbiphenyl-3-yl)methyl ester
- Molecular weight:422,87
- Flash point:Not applicable
- GHS signal: GHS06, GHS08, GHS09
: P202 - P273 - P280 - P301 + P310 - P302 + P352 - P304 + P340 + P311