Supelco® PESTANAL® Organic Reference Standard, Vinclozolin
Catalog # 45705-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:50471-44-8
- Description:Vinclozolin
- Storage conditions:2...30 °C
- Hazard class description:H317 - H351 - H360FD - H411
- Shipping conditions:STD
- MDL No.:MFCD00055511
- M-Clarity quality level:MQ100
- Molecular weight:286,11
Specifications
About this item
- GHS signal: GHS08, GHS07, GHS09
Caution: P201 - P273 - P280 - P302 + P352 - P308 + P313