Pesticide reference standard, Bifenthrin 100 µg/ml in acetonitrile, PESTANAL®, Supelco®
36993-2ML
: Merck
:
- Pack type:Glass bottle
- Pk:2 mL
- CAS No.:82657-04-3
- Description:Bifenthrin 100 µg/ml in acetonitrile
- M-Clarity quality level:MQ100
- Formula:C₂₃H₂₂ClF₃O₂
- Description:Bifenthrin