Supelco® PESTANAL® Organic Reference Standard, Permethrin
45614-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:52645-53-1
- Description:Permethrin
- Storage conditions:2...30 °C
- Hazard class description:H302 + H332 - H317 - H410
- Shipping conditions:STD
- MDL No.:MFCD00041809
- M-Clarity quality level:MQ100
- Molecular weight:391,29
- Flash point:Not applicable
- GHS signal: GHS07, GHS09
: P261 - P273 - P280 - P301 + P312 - P302 + P352 - P304 + P340 + P312