Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Amitraz
  45323-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Amitraz
  45323-250MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    33089-61-1
  • Description:
    Amitraz
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H302 - H317 - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00069396
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N′-(2,4-Dimethylphenyl)-N-{[(2,4-dimethylphenyl)imino]methyl}-N-methylmethanimidamide; NSC 324552
  • Molecular weight:
    293,41
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS08, GHS07, GHS09
 : P260 - P273 - P280 - P301 + P312 - P302 + P352 - P314