Supelco® PESTANAL® Organic Reference Standard, 2-Imidazolidone
Catalog # 31534-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:120-93-4
- Description:2-Imidazolidone
- Storage conditions:2...30 °C
- Hazard class description:H319 - H373
- Shipping conditions:STD
- MDL No.:MFCD00005257
- M-Clarity quality level:MQ100
- Synonym(s):N,N′-Ethyleneurea
- Melting point:129...132 °C (lit.)
- Molecular weight:86,09
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P260 - P264 - P280 - P305 + P351 + P338 - P314 - P337 + P313