Supelco® PESTANAL® Organic Reference Standard, Propanil
45639-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:709-98-8
- Description:Propanil
- Storage conditions:2...30 °C
- Hazard class description:H302 - H400
- Shipping conditions:STD
- MDL No.:MFCD00018149
- M-Clarity quality level:MQ100
- Synonym(s):3′,4′-Dichloropropionanilide
- Molecular weight:218,08
- Flash point:Not applicable
- GHS signal: GHS07, GHS09
: P264 - P270 - P273 - P301 + P312 - P391 - P501